Learn More
Medchemexpress LLC UDP-glucuronic acid trisodium | 63700-19-6 | 250 MG
SDP
Catalog No. 5000420696
Encompass_Preferred
UDP-glucuronic acid trisodium is a vital precursor for essential glycoconjugates found across various biological kingdoms, including mammalian glycosaminoglycans, plant cell wall polysaccharides, and bacterial capsule glycoglycerolipids. Intended for research use only, this white to off-white solid has a high purity of 99.87%, a molecular weight of 646.23, and a chemical formula of C15H19N2Na3O18P2.
  • Critical precursor for glycoconjugates
  • High purity of 99.87%
  • Promotes migration of liver cancer cells (Huh7, SK-Hep1, SNU449) by inducing EMT process in vitro
  • Decreases E-cadherin levels and increases vimentin levels in Huh7 and SNU449 cells at 0.5 mM in vitro
Catalog No. 50-004-20696 Supplier Medchemexpress LLC Supplier No. 250MGHYN7033
UDP-glucuronic acid trisodium is a vital precursor for essential glycoconjugates found across various biological kingdoms, including mammalian glycosaminoglycans, plant cell wall polysaccharides, and bacterial capsule glycoglycerolipids. Intended for research use only, this white to off-white solid has a high purity of 99.87%, a molecular weight of 646.23, and a chemical formula of C15H19N2Na3O18P2.
  • Critical precursor for glycoconjugates
  • High purity of 99.87%
  • Promotes migration of liver cancer cells (Huh7, SK-Hep1, SNU449) by inducing EMT process in vitro
  • Decreases E-cadherin levels and increases vimentin levels in Huh7 and SNU449 cells at 0.5 mM in vitro