Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC UDP-glucuronic acid (trisodium) | 63700-19-6 | 50 MG
SDP

Catalog No. 5000418987
Encompass_Preferred

UDP-glucuronic acid trisodium (Uridine-5'-diphosphoglucuronic acid trisodium salt) is a critical precursor for essential glycoconjugates across biological kingdoms, ranging from mammalian glycosaminoglycans and plant cell wall polysaccharides to bacterial capsule glycoglycerolipids.

  • Purity of 99.87%
  • Molecular weight of 646.23
  • Chemical formula: C15H19N2Na3O18P2
  • Appears as a white to off-white solid
  • Soluble in H2O and DMSO
  • Melting point above 175°C
  • Store at -20°C, sealed and away from moisture

Catalog No. 50-004-18987 Supplier Medchemexpress LLC Supplier No. HYN703350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

UDP-glucuronic acid trisodium (Uridine-5'-diphosphoglucuronic acid trisodium salt) is a critical precursor for essential glycoconjugates across biological kingdoms, ranging from mammalian glycosaminoglycans and plant cell wall polysaccharides to bacterial capsule glycoglycerolipids.

  • Purity of 99.87%
  • Molecular weight of 646.23
  • Chemical formula: C15H19N2Na3O18P2
  • Appears as a white to off-white solid
  • Soluble in H2O and DMSO
  • Melting point above 175°C
  • Store at -20°C, sealed and away from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.