Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC UDP-glucuronic acid (trisodium) | 63700-19-6 | 99.9% | 646.23 | 10 MG
SDP

Catalog No. 5000365144
Encompass_Preferred

UDP-glucuronic acid trisodium (Uridine-5'-diphosphoglucuronic acid trisodium salt) is a critical precursor for essential glycoconjugates across biological kingdoms, including mammalian glycosaminoglycans, plant cell wall polysaccharides, and bacterial capsule glycoglycerolipids. This product is a laboratory chemical intended for research use only by qualified scientists.

  • Precursor for essential glycoconjugates in various biological organisms
  • Promotes migration of liver cancer cells (Huh7, SK-Hep1, SNU449) by inducing the EMT process in vitro
  • Decreases E-cadherin levels and increases vimentin levels in Huh7 and SNU449 cells in vitro
  • Soluble in H2O (≥ 100 mg/mL) and DMSO (20 mg/mL with warming/ultrasonic)
  • Recommended storage at -20°C, sealed, away from moisture
  • In solvent, recommended storage at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-65144 Supplier Medchemexpress LLC Supplier No. 10MGHYN7033
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

UDP-glucuronic acid trisodium (Uridine-5'-diphosphoglucuronic acid trisodium salt) is a critical precursor for essential glycoconjugates across biological kingdoms, including mammalian glycosaminoglycans, plant cell wall polysaccharides, and bacterial capsule glycoglycerolipids. This product is a laboratory chemical intended for research use only by qualified scientists.

  • Precursor for essential glycoconjugates in various biological organisms
  • Promotes migration of liver cancer cells (Huh7, SK-Hep1, SNU449) by inducing the EMT process in vitro
  • Decreases E-cadherin levels and increases vimentin levels in Huh7 and SNU449 cells in vitro
  • Soluble in H2O (≥ 100 mg/mL) and DMSO (20 mg/mL with warming/ultrasonic)
  • Recommended storage at -20°C, sealed, away from moisture
  • In solvent, recommended storage at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.