Learn More
Carbosynth LLC. UDP-β-L-Rhamnose (0.1 mg vial), CAS# 1955-26-6, MF= C15H24N2O16P2
SDP
Supplier:  Carbosynth LLC. CKF23009S0.1MG
Encompass_Preferred
Uridine 5'-(β-L-Rhamnopyranosyl Diphosphate); UDP-β-L-Rha, Application: Chemically Synthesized Nucleotide Sugar as a Glycosyl Donor for Glycosyltransferases in Plants, salt: Sodium Salt, Synonym: UDP-β-L-Rha, MW: 550.3
Catalog No. 50-195-2402