Learn More
Medchemexpress LLC UDP-N-azidoacetylgalactosamine disodium | 653600-61-4 | 99.9% | 692.33 | C17H24N6Na2O17P2 | 5 MG
SDP
Catalog No. 5000221582
Encompass_Preferred
UDP-GalNAz disodium is an azide-bearing nucleotide sugar analogue (UDP-N-azidoacetylgalactosamine disodium) used as a donor substrate for N-acetylgalactosaminyltransferases and as a bioorthogonal reagent for azide-alkyne click chemistry. Supplied as the water-soluble disodium salt, it is intended for in vitro glycosylation, biochemical assays, and click-conjugation workflows requiring an azide-functionalized UDP-sugar.
Catalog No. 50-002-21582 Supplier Medchemexpress LLC Supplier No. HY1459345MG
UDP-GalNAz disodium is an azide-bearing nucleotide sugar analogue (UDP-N-azidoacetylgalactosamine disodium) used as a donor substrate for N-acetylgalactosaminyltransferases and as a bioorthogonal reagent for azide-alkyne click chemistry. Supplied as the water-soluble disodium salt, it is intended for in vitro glycosylation, biochemical assays, and click-conjugation workflows requiring an azide-functionalized UDP-sugar.