Learn More
Medchemexpress LLC UDP-rhamnose | 1955-26-6 | 550.30 | 5 MG
SDP
Catalog No. 5000416568
Encompass_Preferred
UDP-rhamnose is the activated form of Rhamnose found in fungi and serves as a crucial precursor for fungi to synthesize rhamnose-containing glycans. This compound is produced through the action of dual enzymes and can be utilized in research focused on treating fungal diseases. It is involved in various interactions between fungi and hosts, including adhesion, recognition, virulence, and biofilm formation.
  • Key precursor for fungi to synthesize rhamnose-containing glycans.
  • Useful in research on the treatment of fungal diseases.
  • Involved in fungal-host interactions like adhesion, recognition, virulence, and biofilm formation.
  • Purity: 96.47%.
Catalog No. 50-004-16568 Supplier Medchemexpress LLC Supplier No. HYN105735MG
UDP-rhamnose is the activated form of Rhamnose found in fungi and serves as a crucial precursor for fungi to synthesize rhamnose-containing glycans. This compound is produced through the action of dual enzymes and can be utilized in research focused on treating fungal diseases. It is involved in various interactions between fungi and hosts, including adhesion, recognition, virulence, and biofilm formation.
  • Key precursor for fungi to synthesize rhamnose-containing glycans.
  • Useful in research on the treatment of fungal diseases.
  • Involved in fungal-host interactions like adhesion, recognition, virulence, and biofilm formation.
  • Purity: 96.47%.