Learn More
Medchemexpress LLC UDP-xylose (disodium) | 108320-89-4 | 580.24 | 100 MG
SDP
Catalog No. 5000414828
Encompass_Preferred
UDP-xylose disodium is an endogenous sugar nucleotide that functions as a catalytic substrate of UDP-xylose disodium synthase (UXS). It acts as a sugar donor in the synthesis of various biological molecules and is involved in the regulation of extracellular matrix components.
  • Functions as a sugar donor for glycoproteins, polysaccharides, metabolites, and oligosaccharides
  • Participates in proteoglycan synthesis as a glycosyl donor
  • Regulates synthesis of extracellular matrix components
  • Useful for studying proteoglycan biosynthesis in glycobiology and related diseases
Catalog No. 50-004-14828 Supplier Medchemexpress LLC Supplier No. HYN11287A100MG
UDP-xylose disodium is an endogenous sugar nucleotide that functions as a catalytic substrate of UDP-xylose disodium synthase (UXS). It acts as a sugar donor in the synthesis of various biological molecules and is involved in the regulation of extracellular matrix components.
  • Functions as a sugar donor for glycoproteins, polysaccharides, metabolites, and oligosaccharides
  • Participates in proteoglycan synthesis as a glycosyl donor
  • Regulates synthesis of extracellular matrix components
  • Useful for studying proteoglycan biosynthesis in glycobiology and related diseases