Learn More
Medchemexpress LLC UDP-xylose disodium | 108320-89-4 | 580.24 | 5 MG
SDP
Catalog No. 5000416354
Encompass_Preferred
UDP-xylose disodium is an endogenous sugar nucleotide and a catalytic substrate of UDP-xylose disodium synthase (UXS). It serves as a crucial sugar donor for the synthesis of various biomolecules, including glycoproteins, polysaccharides, and oligosaccharides, across plants, vertebrates, and fungi. This compound also plays a vital role in proteoglycan synthesis as a glycosyl donor and is involved in regulating the synthesis of extracellular matrix components.
  • Endogenous sugar nucleotide
  • Catalytic substrate of UDP-xylose disodium synthase (UXS)
  • Sugar donor for synthesis of glycoproteins, polysaccharides, and oligosaccharides
  • Participates in proteoglycan synthesis
  • Regulates extracellular matrix components synthesis
  • Useful for studying proteoglycan biosynthesis in glycobiology
  • Aids research into related diseases like connective tissue diseases
Catalog No. 50-004-16354 Supplier Medchemexpress LLC Supplier No. HYN11287A5MG
UDP-xylose disodium is an endogenous sugar nucleotide and a catalytic substrate of UDP-xylose disodium synthase (UXS). It serves as a crucial sugar donor for the synthesis of various biomolecules, including glycoproteins, polysaccharides, and oligosaccharides, across plants, vertebrates, and fungi. This compound also plays a vital role in proteoglycan synthesis as a glycosyl donor and is involved in regulating the synthesis of extracellular matrix components.
  • Endogenous sugar nucleotide
  • Catalytic substrate of UDP-xylose disodium synthase (UXS)
  • Sugar donor for synthesis of glycoproteins, polysaccharides, and oligosaccharides
  • Participates in proteoglycan synthesis
  • Regulates extracellular matrix components synthesis
  • Useful for studying proteoglycan biosynthesis in glycobiology
  • Aids research into related diseases like connective tissue diseases