Learn More
Cayman Chemical UDP-α-D-Galactose (sodium salt), Purity: ≥95%, CAS: 137868-52-1, 10 mg
SDP
Catalog No. 502950063
Encompass_Preferred
A nucleotide sugar; an agonist of the purinergic P2Y14 receptor (EC50 = 0.67 µM); red blood cell levels are decreased in patients with PKU and maple syrup urine disease
Catalog No. 50-295-0063 Supplier Cayman Chemical Supplier No. 3466410 MG
A nucleotide sugar; an agonist of the purinergic P2Y14 receptor (EC50 = 0.67 µM); red blood cell levels are decreased in patients with PKU and maple syrup urine disease