Learn More
Cayman Chemical UDPaDGalactosesodIM sal 50mg
SDP
Catalog No. 502946133
Encompass_Preferred
A nucleotide sugar; an agonist of the purinergic P2Y14 receptor (EC50 = 0.67 µM); red blood cell levels are decreased in patients with PKU and maple syrup urine disease
Catalog No. 50-294-6133 Supplier Cayman Chemical Supplier No. 3466450 MG
A nucleotide sugar; an agonist of the purinergic P2Y14 receptor (EC50 = 0.67 µM); red blood cell levels are decreased in patients with PKU and maple syrup urine disease