Learn More
Cayman Chemical UDPaDGlucosesodIM salt 100mg
SDP
Catalog No. 502972632
Encompass_Preferred
An endogenous nucleotide sugar that binds the P2Y14 receptor (EC50 = 0.35 μM) as well as GPR17; induces oligodendrocyte differentiation at a maximal concentration of 100 μM
Catalog No. 50-297-2632 Supplier Cayman Chemical Supplier No. 15602100 MG
An endogenous nucleotide sugar that binds the P2Y14 receptor (EC50 = 0.35 μM) as well as GPR17; induces oligodendrocyte differentiation at a maximal concentration of 100 μM