Learn More
Cayman Chemical UDP-β-D-Glucose (sodium salt), Purity: ≥95%, CAS: 7333-33-7, 500 mg
SDP
Catalog No. 502986046
Encompass_Preferred
A nucleotide sugar and P2Y14 receptor agonist; induces inositol phosphate production in COS-7 cells expressing the human P2Y14 receptor (EC50 = 0.588 µM)
Catalog No. 50-298-6046 Supplier Cayman Chemical Supplier No. 21620500 MG
A nucleotide sugar and P2Y14 receptor agonist; induces inositol phosphate production in COS-7 cells expressing the human P2Y14 receptor (EC50 = 0.588 µM)