Learn More
Cayman Chemical UDPGlucuronIc AcIdodIum s 10mg
SDP
Catalog No. 502984561
Encompass_Preferred
A source of glucuronic acid for polysaccharide biosynthesis; used for the formation of many glucuronides with various aglycones
Catalog No. 50-298-4561 Supplier Cayman Chemical Supplier No. 2067410 MG
A source of glucuronic acid for polysaccharide biosynthesis; used for the formation of many glucuronides with various aglycones