Learn More
Cayman Chemical UDPNacetylDGlucosamIns 10mg
SDP
Catalog No. 502983848
Encompass_Preferred
A nucleotide sugar that is used by glycosyltransferases to transfer N-acetylglucosamine residues to substrates
Catalog No. 50-298-3848 Supplier Cayman Chemical Supplier No. 2035310 MG
A nucleotide sugar that is used by glycosyltransferases to transfer N-acetylglucosamine residues to substrates