Learn More
Cayman Chemical UDPNacetylDGlucosamIns 50mg
SDP
Catalog No. 502981958
Encompass_Preferred
A nucleotide sugar that is used by glycosyltransferases to transfer N-acetylglucosamine residues to substrates
Catalog No. 50-298-1958 Supplier Cayman Chemical Supplier No. 2035350 MG
A nucleotide sugar that is used by glycosyltransferases to transfer N-acetylglucosamine residues to substrates