Learn More
Cayman Chemical UDP-β-S (sodium salt), Purity: ≥95%, 2.1 mg
SDP
Catalog No. 502917244
Encompass_Preferred
A P2Y6 receptor agonist; selectively induces inositol phosphate accumulation in 1321N1 cells expressing P2Y6 receptors (EC50 = 28 nM) over those expressing P2Y1, P2Y2, or P2Y4receptors at 10 µM; induces DNA synthesis in isolated rat aorta smooth muscle cells at 1 µM
Catalog No. 50-291-7244 Supplier Cayman Chemical Supplier No. 384932.1 MG
A P2Y6 receptor agonist; selectively induces inositol phosphate accumulation in 1321N1 cells expressing P2Y6 receptors (EC50 = 28 nM) over those expressing P2Y1, P2Y2, or P2Y4receptors at 10 µM; induces DNA synthesis in isolated rat aorta smooth muscle cells at 1 µM