Learn More
Cayman Chemical UDPsodIum salt hydrate 100mg
SDP
Catalog No. 502965482
Encompass_Preferred
The diphosphate of uridine; a specific agonist of the P2Y purinoceptor P2Y6 (EC50 = 13 nM for human P2Y6), stimulating the production of inflammatory mediators, phagocytosis, and vasoconstriction; an antagonist of P2Y14
Catalog No. 50-296-5482 Supplier Cayman Chemical Supplier No. 18137100 MG
The diphosphate of uridine; a specific agonist of the P2Y purinoceptor P2Y6 (EC50 = 13 nM for human P2Y6), stimulating the production of inflammatory mediators, phagocytosis, and vasoconstriction; an antagonist of P2Y14