Learn More
Cayman Chemical UDPsodIum salt hydrate 500mg
SDP
Catalog No. 502964946
Encompass_Preferred
The diphosphate of uridine; a specific agonist of the P2Y purinoceptor P2Y6 (EC50 = 13 nM for human P2Y6), stimulating the production of inflammatory mediators, phagocytosis, and vasoconstriction; an antagonist of P2Y14
Catalog No. 50-296-4946 Supplier Cayman Chemical Supplier No. 18137500 MG
The diphosphate of uridine; a specific agonist of the P2Y purinoceptor P2Y6 (EC50 = 13 nM for human P2Y6), stimulating the production of inflammatory mediators, phagocytosis, and vasoconstriction; an antagonist of P2Y14