Learn More
Vector Laboratories Succinylated WGA, Biotinylated DL-1067-1
SDP
Catalog No. 502045645
Encompass
Succinylated Wheat Germ Agglutinin (WGA), Biotinylated, 5 mg. This derivative has been reported to have properties distinct from the native lectin. Evidence suggests that Succinylated Wheat Germ agglutinin does not bind to sialic acid residues, unlike the native form, but retains its specificity toward N-acetylglucosamine. Biotinylated, succinylated WGA has an appropriate number of biotins bound to provide the optimum staining characteristics for this lectin. This conjugate is supplied essentially free of unconjugated biotins and is preserved with sodium azide.
Catalog No. 50-204-5645 Supplier Vector Laboratories Supplier No. DL10671
Add to Cart
Add to Cart
Succinylated Wheat Germ Agglutinin (WGA), Biotinylated, 5 mg. This derivative has been reported to have properties distinct from the native lectin. Evidence suggests that Succinylated Wheat Germ agglutinin does not bind to sialic acid residues, unlike the native form, but retains its specificity toward N-acetylglucosamine. Biotinylated, succinylated WGA has an appropriate number of biotins bound to provide the optimum staining characteristics for this lectin. This conjugate is supplied essentially free of unconjugated biotins and is preserved with sodium azide.