Learn More
Vector Laboratories Ulex Europaeus Agglutinin I (UEA I) Fluorescein-labeled 5 mg
SDP
Catalog No. NC0776461
Encompass
Ulex europaeus agglutinin I (UAE I) binds to many glycoproteins and glycolipids containing α-linked fucose residues, such as ABO blood group glycoconjugates. This lectin preferentially binds blood group O cells and has been used to determine secretor status. It is also an established marker for human endothelial cells. This fluorescein-labeled UAE I features a ratio of fluorophores to lectin protein that provides optimal staining (excitation 495 nm, emission 515 nm). Supplied as a solution essentially free of unconjugated fluorophores, it is preserved with sodium azide. The recommended inhibiting/eluting sugar is 50-100 mM L-fucose.
Catalog No. NC0776461 Supplier Vector Laboratories Supplier No. FL10615
Ulex europaeus agglutinin I (UAE I) binds to many glycoproteins and glycolipids containing α-linked fucose residues, such as ABO blood group glycoconjugates. This lectin preferentially binds blood group O cells and has been used to determine secretor status. It is also an established marker for human endothelial cells. This fluorescein-labeled UAE I features a ratio of fluorophores to lectin protein that provides optimal staining (excitation 495 nm, emission 515 nm). Supplied as a solution essentially free of unconjugated fluorophores, it is preserved with sodium azide. The recommended inhibiting/eluting sugar is 50-100 mM L-fucose.