Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ufc1 protein, human (sf9, His, StrepⅡ) | >90.0% | 10UG
SDP

Catalog No. 5000176071
Encompass_Preferred

Recombinant human UFC1 protein expressed in Sf9 insect cells and supplied as a purified, buffered solution for research applications. The construct contains N-terminal StrepII and 8xHis tags, has an approximate molecular weight of 22.5 kDa, and is provided with low endotoxin and high purity. Store frozen and avoid repeated freeze-thaw cycles.

  • Recombinant human UFC1 protein expressed in Sf9 insect cells.
  • N-terminal StrepII and 8xHis tags for purification and detection.
  • Approximate molecular weight 22.5 kDa.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Supplied in 50 mM HEPES pH 7.5, 200 mM NaCl, 20% glycerol, 1 mM DTT.
  • Endotoxin level <1 EU/μg (LAL method).
  • Shipped on dry ice; store at -80°C for long-term storage.
  • Intended for research use only.

Catalog No. 50-001-76071 Supplier Medchemexpress LLC Supplier No. HYP70149210UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human UFC1 protein expressed in Sf9 insect cells and supplied as a purified, buffered solution for research applications. The construct contains N-terminal StrepII and 8xHis tags, has an approximate molecular weight of 22.5 kDa, and is provided with low endotoxin and high purity. Store frozen and avoid repeated freeze-thaw cycles.

  • Recombinant human UFC1 protein expressed in Sf9 insect cells.
  • N-terminal StrepII and 8xHis tags for purification and detection.
  • Approximate molecular weight 22.5 kDa.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Supplied in 50 mM HEPES pH 7.5, 200 mM NaCl, 20% glycerol, 1 mM DTT.
  • Endotoxin level <1 EU/μg (LAL method).
  • Shipped on dry ice; store at -80°C for long-term storage.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.