Learn More
Aviva Systems Biology Corp UFD1 Antibody - middle region (ARP59424_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59424_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UFD1L. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human UFD1L. Alternate names: UFD1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-691