Learn More
Aviva Systems Biology Corp UFM1 Antibody - middle region (ARP59400_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59400_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UFM1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human UFM1. Alternate names: BM-002, C13orf20, bA131P10.1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-687