Learn More
ABclonal Technology UFM1 Rabbit mAb
SDP
Catalog No. 5000008172
Encompass_Preferred
UFM1 is a ubiquitin-like protein that is conjugated to target proteins by E1-like activating enzyme UBA5 (UBE1DC1 MIM 610552) and E2-like conjugating enzyme UFC1 (MIM 610554) in a manner analogous to ubiquitylation (see UBE2M MIM 603173) (Komatsu et al 2004 [PubMed 15071506])
Catalog No. 50-000-08172 Supplier ABclonal Technology Supplier No. A2087520UL
UFM1 is a ubiquitin-like protein that is conjugated to target proteins by E1-like activating enzyme UBA5 (UBE1DC1 MIM 610552) and E2-like conjugating enzyme UFC1 (MIM 610554) in a manner analogous to ubiquitylation (see UBE2M MIM 603173) (Komatsu et al 2004 [PubMed 15071506])