Learn More
Cell Signaling Technology UFMylation Antibody Sampler Kit, Unconjugated, 1 Kit
SDP
Catalog No. 502732412
Encompass
The UFMylation Antibody Sampler Kit provides an economical solution for detecting proteins associated with UFMylation during ER-phagy. This kit includes a selection of rabbit monoclonal antibodies targeting key components and substrates of the ubiquitin-fold modifier 1 (UFM1) conjugation system, including UFL1, UFM1, DDRGK1, and Ribosomal Protein L26, along with an HRP-linked secondary antibody.
  • Designed for the detection of UFMylation components and substrates involved in ER homeostasis and autophagy.
  • Includes sufficient antibody volume to perform two western blot experiments with each primary antibody.
  • Validated for multiple applications including western blotting, immunofluorescence, and immunoprecipitation (agarose and magnetic).
  • Supplied in a stabilized glycerol-based storage buffer for convenient long-term use at −20°C.
Catalog No. 50-273-2412 Supplier Cell Signaling Technology Supplier No. 63522T
The UFMylation Antibody Sampler Kit provides an economical solution for detecting proteins associated with UFMylation during ER-phagy. This kit includes a selection of rabbit monoclonal antibodies targeting key components and substrates of the ubiquitin-fold modifier 1 (UFM1) conjugation system, including UFL1, UFM1, DDRGK1, and Ribosomal Protein L26, along with an HRP-linked secondary antibody.
  • Designed for the detection of UFMylation components and substrates involved in ER homeostasis and autophagy.
  • Includes sufficient antibody volume to perform two western blot experiments with each primary antibody.
  • Validated for multiple applications including western blotting, immunofluorescence, and immunoprecipitation (agarose and magnetic).
  • Supplied in a stabilized glycerol-based storage buffer for convenient long-term use at −20°C.