Learn More
Medchemexpress LLC HY-P1299A 1mg Medchemexpress, UFP-101 (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1299A1MG
Encompass_Preferred
Medchemexpress, HY-P1299A 1mg UFP-101 (TFA) CAS: UFP-101 TFA is a potent, selective, and competitive antagonist of the NOP receptor, with a pKi of 10.24. UFP-101 TFA displays >3000-fold selectivity over δ, μ and κ opioid receptors. UFP-101 TFA shows antidepressant-like effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3049