Learn More
Aviva Systems Biology Corp UGCGL1 Antibody - middle region (ARP49467_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49467_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UGCGL1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human UGCGL1. Alternate names: FLJ23671, FLJ23796, HUGT1, UGT1, UGCGL1ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-285-428