Learn More
Aviva Systems Biology Corp UGCGL2 Antibody - N-terminal region (ARP48851_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48851_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UGCGL2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human UGCGL2. Alternate names: FLJ10873, FLJ11485, HUGT2, MGC117360, MGC150689, MGC87276, UGT2, UGCGL2ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-59939