Learn More
Aviva Systems Biology Corp UGCGL2 Antibody - middle region (ARP48852_P050) | 100uL
SDP
Catalog No. 5001459940
Encompass_Preferred
This is a rabbit polyclonal antibody against UGCGL2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human UGCGL2. Alternate names: FLJ10873, FLJ11485, HUGT2, MGC117360, MGC150689, MGC87276, UGT2, UGCGL2ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-59940 Supplier Aviva Systems Biology Corp Supplier No. ARP48852_P050
This is a rabbit polyclonal antibody against UGCGL2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human UGCGL2. Alternate names: FLJ10873, FLJ11485, HUGT2, MGC117360, MGC150689, MGC87276, UGT2, UGCGL2ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast