Learn More
Aviva Systems Biology Corp UGDH Antibody - C-terminal region (ARP61247_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61247_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UGDH. It was validated on Western Blot. . Alternate names: GDH, UDP-GlcDH, UDPGDH, UGDApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-312