Learn More
Stoelting™ Ugo Basile Active Avoidance, Mouse
SDP
Catalog No. 10000949 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Mouse
Catalog No. Animal
10-000-949 Mouse
1 options
Catalog No. 10-000-949 Supplier Stoelting™ Supplier No. 55533
Add to Cart
Add to Cart
Request a Quote
Stoelting™ Ugo Basile Active Avoidance (Shuttle-Box) Apparatus is a fear-motivated task in which the animal learns to predict the occurrence of an aversive event (shock) based on the presence of a stimulus (tone) and moves to a second compartment to avoid the aversive event (shock).
Specifications
For Use With (Application) Neuroscience
Animal Mouse