Learn More
Stoelting™ Ugo Basile Active Avoidance
SDP
Catalog No. 10000948 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Mouse
Catalog No. Animal
10-000-948 Mouse
1 options
Catalog No. 10-000-948 Supplier Stoelting™ Supplier No. 55532
Add to Cart
Add to Cart
Request a Quote
Stoelting™ Ugo Basile Active Avoidance (Shuttle-Box) Apparatus is a fear-motivated task in which the animal learns to predict the occurrence of an aversive event (shock) based on the presence of a stimulus (tone) and moves to a second compartment to avoid the aversive event (shock).
Specifications
For Use With (Application) Neuroscience
Animal Mouse