Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stoelting™ Ugo Basile Activity Cage
SDP

Catalog No. 10000989 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Activity cage
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-000-989 Activity cage
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-000-989 Supplier Stoelting™ Supplier No. 57445
Add to Cart
Add to Cart
Request a Quote

Stoelting™ Ugo Basile Activity Cage measures horizontal and vertical locomotor activity in mice and rats.

Specifications

For Use With (Application) Neuroscience
Model Activity cage
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.