Learn More
Stoelting™ Ugo Basile Dynamic Plantar Aesthesiometer
SDP
Catalog No. 10001001 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Mouse, Rat
Catalog No. Animal
10-001-001 Mouse, Rat
1 options
Catalog No. 10-001-001 Supplier Stoelting™ Supplier No. 57820
Add to Cart
Add to Cart
Request a Quote
Stoelting™ Ugo Basile Dynamic Plantar Aesthesiometer has been designed to automate the assessment of touch sensitivity on the plantar surface of rats or mice
  • Comes ready to work with both mice and rats
  • Differs from Von Frey devices as Latency and Actual Force at the time of paw withdrawal reflex are automatically detected and recorded
Specifications
For Use With (Application) Neuroscience
Animal Mouse, Rat