Learn More
Stoelting™ Ugo Basile Electronic Von Frey with Grid Platform
SDP
Catalog No. 10001000 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Mouse, Rat
Catalog No. Animal
10-001-000 Mouse, Rat
1 options
Catalog No. 10-001-000 Supplier Stoelting™ Supplier No. 57815
Add to Cart
Add to Cart
Request a Quote
Stoelting™ Ugo Basile Electronic von Frey instrument provides automated assessment of theshold to mechanical stimuli in mice and rats.  
  • Designed for applying light touch to the rodent foot
  • Provides assessment of hypersensitivity and allodynia in rats and mice
  • Touch stimulator transducer is mounted on a Perspex handle, so that routine procedures may be employed to examine and test the animal skin sensitivity
  • Prism of original design is of great help to localize the aimed stimulation area
  • Completion of each test may be indicated either by the sudden withdrawal of the paw or by pressing the external foot-pedal
  • Display gives the operator a summary of the results of the test (i.e. force and time corresponding to the animal response).
  • Rate of application of the force is set by the operator and the e-VF includes software tools that help the operator in consistently applying the force at the desired rate.
  • Results of several hundred tests may be stored in the e-VF for transfer to a PC
Instrument includes:
  • Electronic unit
  • Touch stimulator with the prism of original design 
  • e-VF can be completed with the 57816 Base assembly for plantar stimulation.
Specifications
For Use With (Application) Neuroscience
Animal Mouse, Rat
Includes Grid platform