Learn More
Stoelting™ Ugo Basile Learned Helplessness Apparatus, Mouse
SDP
Catalog No. 10000991 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Mouse
Catalog No. Animal
10-000-991 Mouse
1 options
Catalog No. 10-000-991 Supplier Stoelting™ Supplier No. 57501
Add to Cart
Add to Cart
Request a Quote
Stoelting™ Ugo Basile Learned-Helplessness Apparatus is widely used for assessing depression in rats and mice. When rodents are exposed to unavoidable and unpredictable stress, they often develop learning, memory and other behavioral deficits and show analgesic reactions
Specifications
For Use With (Application) Neuroscience
Animal Mouse