Learn More
Stoelting™ Ugo Basile Lesion Making Device
SDP
Catalog No. 10000927 Shop All Stoelting Co Products
Change view
Click to view available options
For Use With (Application):
Neuroscience
Catalog No. For Use With (Application)
10-000-927 Neuroscience
1 options
Catalog No. 10-000-927 Supplier Stoelting™ Supplier No. 53500
Add to Cart
Add to Cart
Request a Quote
Stoelting™ Ugo Basile Lesion Making Device is a compact, solid state direct current lesion maker designed for the production of localized lesions in small animals.
Current generator is protected against short circuit, which prevents the electronics from getting damaged when the electrodes accidentally come into contact with each other.
Specifications
For Use With (Application) Neuroscience