Learn More
Stoelting™ Ugo Basile Multi-Maze
SDP
Catalog No. 10001103 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Mouse
Rat
Model:
Central arena
Standard arm with automated door
Start compartment w/automated door
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Animal Model
10-001-103 Rat Start compartment w/automated door
10-001-098 Mouse Central arena
10-001-099 Mouse Standard arm with automated door
10-001-100 Mouse Start compartment w/automated door
10-001-101 Rat Central arena
10-001-102 Rat Standard arm with automated door
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 10-001-103 Supplier Stoelting™ Supplier No. 675063
Add to Cart
Add to Cart
Request a Quote
Stoelting™ Ugo Basile Multi-Maze for Mice is a modular system providing full automation of Y-maze, T-maze and 8-arm Radial Arm Maze.
Allows for conducting fully automated memory experiments including:
  • Assessment of spatial memory
  • Basic working memory
  • Discriminating working from reference memory
  • Evaluating impairments in working memory
Includes a central electronic unit and up to eight modular units that can be configured as:
  • Y-maze
  • T-maze
  • 8-arm Radial Arm Maze
Proprietary sliding doors retract into the maze floor smoothly and silently without disrupting the animal during testing. This sliding mechanism combined with the non-reflective, high contrast maze material provides optimal results with ANY-maze Video Tracking Software (sold separately).
Specifications
For Use With (Application) Neuroscience
Animal Rat
Model Start compartment w/automated door