Learn More
Stoelting™ Ugo Basile Passive Avoidance Apparatus
SDP
Catalog No. 10000992 Shop All Stoelting Co Products
Change view
Click to view available options
For Use With (Application):
Neuroscience
Catalog No. For Use With (Application)
10-000-992 Neuroscience
1 options
Catalog No. 10-000-992 Supplier Stoelting™ Supplier No. 57550
Add to Cart
Add to Cart
Request a Quote
Stoelting™ Ugo Basile Passive Avoidance Apparatus is a classic instrument for Pavlovian Conditioning exploits the tendency in mice and rats to escape from an illuminated area into a dark one (step-through method).
  • The instrument consists of a tilting-floor box (for mice or rats) divided into two compartments by a sliding door and a control unit incorporating a scrambler shocker.
  • Unique elevator door design opens the passage to the dark side after a preset delay
  • Doors operate silently and automatically
  • Latency to pass through the doors is recorded with 0.1 sec. resolution
  • Ccontroller allows digital setting of the shock intensity in milliamps
Specifications
For Use With (Application) Neuroscience