Learn More
Stoelting™ Ugo Basile Passive Avoidance Step-Down Apparatus
SDP
Catalog No. 10000994 Shop All Stoelting Co Products
Change view
Click to view available options
For Use With (Application):
Neuroscience
Catalog No. For Use With (Application)
10-000-994 Neuroscience
1 options
Catalog No. 10-000-994 Supplier Stoelting™ Supplier No. 57560
Add to Cart
Add to Cart
Request a Quote
Stoelting™ Ugo Basile Passive Avoidance Step-Down Apparatus is used for evaluating learning and memory.
Step-down apparatus consists of:
  • Perspex box (28 x 23 x 26 cm)
  • Electrified cage floor (40, 0.2 cm stainless steel bars spaced 0.5 cm apart)
  • Elevated vibrating platform 7 cm
Mice are placed on the elevated platform and when the animal steps off of the platform, a shock is induced. Latency for the mouse to return to the platform is automatically recorded.
Specifications
For Use With (Application) Neuroscience