Learn More
Stoelting™ Ugo Basile Passive Avoidance (Step-Through), Mouse
SDP
Catalog No. 10000993 Shop All Stoelting Co Products
Change view
Click to view available options
For Use With (Application):
Neuroscience
Catalog No. For Use With (Application)
10-000-993 Neuroscience
1 options
Catalog No. 10-000-993 Supplier Stoelting™ Supplier No. 57553
Add to Cart
Add to Cart
Request a Quote
Stoelting™ Basile Passive Avoidance Instrument is an efficient, reliable instrument for the classic passive avoidance task.
Rodents will normally crawl through a small opening into a darker chamber. If they are shocked in the dark chamber… this normal tendency is reduced, measured by longer latency or refusal to enter the dark chamber …that is, to passively stay where they are.
Specifications
For Use With (Application) Neuroscience