Learn More
Stoelting™ Ugo Basile Paw Pressure Analgesia Meter
SDP
Catalog No. 10000982 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Mouse
Rat
Model:
Meter
Meter, Recording
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Animal Model
10-000-982 Mouse Meter, Recording
10-000-979 Rat Meter
10-000-980 Mouse Meter
10-000-981 Rat Meter, Recording
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 10-000-982 Supplier Stoelting™ Supplier No. 57223
Add to Cart
Add to Cart
Request a Quote
Stoelting™ Ugo Basile Paw Pressure Analgesia Meter is for the rapid precise screening of analgesic drugs
Designed for the Randall-Selitto test, which requires the application of increasing pressure on the paw between a flat surface and a blunt pointer. Usually used with animals with one paw inflamed by an injection and one normal paw to evaluate drugs for analgesic action.Principle of OperationThe device exerts a steadily increasing force (16 grams per second) on the rat paw until the animal makes a stereotyped flinch response. The researcher releases a foot pedal to stop the force indicator. The electronics measures the time interval between the start of the slide motion and the moment the slide is stopped by the operator and converts time into force.The force is measured on the scale calibrated in 10-gram steps, by a pointer riveted to the slide. The scale can be multiplied by 2 or 3, by placing one or two disc weights on the slide.
Specifications
For Use With (Application) Neuroscience
Animal Mouse
Model Meter, Recording