Learn More
Stoelting™ Ugo Basile Plethysmometer
SDP
Catalog No. 10000978 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Mouse, Rat
Catalog No. Animal
10-000-978 Mouse, Rat
1 options
Catalog No. 10-000-978 Supplier Stoelting™ Supplier No. 57140
Add to Cart
Add to Cart
Request a Quote
Stoelting™ Ugo Basile Plethysmometer is designed specifically to measure paw swelling in rodents.
In research on rheumatoid arthritis, the central development of oedema, and its modifications by pharmacological processes, it has proved of great value to measure inflammatory oedema in the rat and mouse paw.
Specifications
For Use With (Application) Neuroscience
Animal Mouse, Rat