Learn More
Stoelting™ Ugo Basile Pressure Application Measurement (PAM)
SDP
Catalog No. 10001010 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
PAM Meter
Catalog No. Model
10-001-010 PAM Meter
1 options
Catalog No. 10-001-010 Supplier Stoelting™ Supplier No. 58500
Add to Cart
Add to Cart
Request a Quote
Stoelting™ Pressure Application Measurement (PAM) device is used for measuring mechanical pain threshold in experimental joint hypersensitivity models in rodents.
Designed and validated specifically for the mechanical stimulation and assessment of joint pain, ideal for detecting arthritis in rats and mice.
Specifications
For Use With (Application) Neuroscience
Animal Mouse, Rat
Model PAM Meter