Learn More
Stoelting™ Ugo Basile Rota-Rod
SDP
Catalog No. 10000995 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Mouse
Mouse, Rat
Rat
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Animal
10-000-995 Mouse
10-000-996 Rat
10-000-997 Mouse, Rat
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 10-000-995 Supplier Stoelting™ Supplier No. 57624
Add to Cart
Add to Cart
Request a Quote
Stoelting™ Ugo Basile Rota-Rod is designed to assess the effects of drugs, brain damage, and diseases on motor coordination and fatigue
When the animal falls off its cylinder section onto the plate below, the plate trips, thereby recording the animal's endurance time in seconds.
Main features, digitally preset by function keys are:
  • Zero noise
  • Angular speed can be preset from 5 to 80 RPM in steps of 1 RPM
  • In the acceleration mode, the speed changes can be preset in 6 second to 10 minute intervals in 6 second steps
  • Multi-step ramp available
  • Reverse rotation can be selected, which takes place at minimum speed, at the end of a programmable acceleration-deceleration sequence
  • Rocking motion can be preset with adjustable angular amplitude, speed, and acceleration
  • Tiltable digital panel shows accrued rotations and time on the rod.
  • Includes software for PC connection, and parameters can be controlled via your PC.
Specifications
For Use With (Application) Neuroscience
Animal Mouse