Learn More
Stoelting™ Ugo Basile Startle/PPI System
SDP
Catalog No. 10001009 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Four-cage system
One-cage system
Two-cage system
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-001-009 Four-cage system
10-001-007 One-cage system
10-001-008 Two-cage system
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 10-001-009 Supplier Stoelting™ Supplier No. 58002
Add to Cart
Add to Cart
Request a Quote
Stoelting™ Ugo Basile Startle/PPI System is used to detect startle response in mice. Each system includes one touch screen and mouse cage. Software activation key sold separately.
Specifications
For Use With (Application) Neuroscience
Animal Mouse
Model Four-cage system