Learn More
Stoelting™ Ugo Basile Tail Flick
SDP
Catalog No. 10000983 Shop All Stoelting Co Products
Change view
Click to view available options
For Use With (Application):
Neuroscience
Catalog No. For Use With (Application)
10-000-983 Neuroscience
1 options
Catalog No. 10-000-983 Supplier Stoelting™ Supplier No. 57360
Add to Cart
Add to Cart
Request a Quote
Stoelting™ Ugo Basile Tail Flick has been designed to accurately measure the nociceptive threshold to infrared heat stimulus on the rat or mouse tail.
Operator starts the stimulus: when the animal feels pain and flicks its tail, a sensor detects it, stops the timer and switches off the bulb. The reaction time of the animal is thus determined and automatically recorded.
Specifications
For Use With (Application) Neuroscience