Learn More
Stoelting™ Ugo Basile Thermal Plantar Test
SDP
Catalog No. 10000947 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Mouse, Rat
Catalog No. Animal
10-000-947 Mouse, Rat
1 options
Catalog No. 10-000-947 Supplier Stoelting™ Supplier No. 55370
Add to Cart
Add to Cart
Request a Quote
Stoelting™ Ugo Basile Thermal Plantar Test Instrument measures response to infrared heat stimulus.
  • Focus the infrared source below the Plantar surface and with the press of a button latency to paw withdrawal and infrared intensity are recorded automatically (manual scoring)
  • Combines the best features of all other methods of measuring pain sensitivity
  • Animal is unrestrained and unhandled during experiments
Specifications
For Use With (Application) Neuroscience
Animal Mouse, Rat