Learn More
Stoelting™ Ugo Basile Treadmill
SDP
Catalog No. 10000999 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Treadmill
Treadmill, tethered rodent
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-000-999 Treadmill, tethered rodent
10-000-998 Treadmill
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-000-999 Supplier Stoelting™ Supplier No. 57633
Add to Cart
Add to Cart
Request a Quote
Stoelting™ Ugo Basile Treadmill is used to study the impact of physical activity on health
  • Adjustable speed and slope
  • Contant, accelerating or custom modes
  • Shock grid is incorporated at the end of the lanes should an aversive stimulus be required.
Specifications
For Use With (Application) Neuroscience
Animal Rodent
Model Treadmill, tethered rodent