Learn More
PHYTO AB INC UGP1/2 Antibody Immunogen:AT5G17310 AT3G03250 Background: UGPase is the major glycosyl donor for polysaccharides in all organisms. 150ug/EA
Supplier:  PHYTO AB INC PHY2522S
Encompass_Preferred
PhytoAB Inc. is a contract research organization and plant antibody R&D based services and products supplier. PhytoAB is privately owned company headquatered in the San Francisco Bay Area of California, USA. PhytoAB specializes in antibody production for plant scientic usage and customized services, focus on the current need of plant antibodies to assist the further investigation of the current booming results from deep genome sequencing and increasing protomics studies. We have specialized R&D and customer services teams to fulfill the needs of botany & plant sciences research and industry world wide with high quality, professional and cutting edge services, and offering complete products usage guidance and suggestions for related problem solutions.
Catalog No. NC2877332